Product Details

SNP ID
rs121908579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218661090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCCCTGGCCCTGGCCCGGAAG[C/G]GTGTCCAACTGGGCCTGGTGGCATT
Phenotype
MIM: 603647 MIM: 616014 MIM: 604083
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCS1L PubMed Links

Gene Details

Gene
BCS1L
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079866.1 449 Missense Mutation CGT,GGT R35G NP_001073335.1
NM_001257342.1 449 Missense Mutation CGT,GGT R35G NP_001244271.1
NM_001257343.1 449 Missense Mutation CGT,GGT R35G NP_001244272.1
NM_001257344.1 449 Missense Mutation CGT,GGT R35G NP_001244273.1
NM_001318836.1 449 Intron NP_001305765.1
NM_001320717.1 449 Missense Mutation CGT,GGT R35G NP_001307646.1
NM_004328.4 449 Missense Mutation CGT,GGT R35G NP_004319.1
XM_005246748.2 449 UTR 5 XP_005246805.1
XM_005246749.3 449 Intron XP_005246806.1
XM_006712678.1 449 Missense Mutation CGT,GGT R35G XP_006712741.1
XM_017004631.1 449 Missense Mutation CGT,GGT R35G XP_016860120.1
XM_017004632.1 449 Missense Mutation CGT,GGT R35G XP_016860121.1
XM_017004633.1 449 UTR 5 XP_016860122.1
XM_017004634.1 449 UTR 5 XP_016860123.1
Gene
RNF25
Gene Name
ring finger protein 25
There are no transcripts associated with this gene.

Gene
ZNF142
Gene Name
zinc finger protein 142
There are no transcripts associated with this gene.

View Full Product Details