Product Details
- SNP ID
-
rs115890861
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219263786 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGAAGAGTTCTAACCTTCAGTCT[G/T]TAGCCAAAGCCAGAGTTAACCAACT
- Phenotype
-
MIM: 191110
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TUBA4A
PubMed Links
Gene Details
- Gene
- TUBA4A
- Gene Name
- tubulin alpha 4a
There are no transcripts associated with this gene.
- Gene
- TUBA4B
- Gene Name
- tubulin alpha 4b
There are no transcripts associated with this gene.
View Full Product Details