Product Details

SNP ID
rs115877951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240715946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGAGCTAAAAAGAGACCAAAT[C/T]GACCTGTGGGTGTGGGTGGGGAGGG
Phenotype
MIM: 601255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF1A PubMed Links

Gene Details

Gene
KIF1A
Gene Name
kinesin family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244008.1 6652 UTR 3 NP_001230937.1
NM_001320705.1 6652 UTR 3 NP_001307634.1
XM_005247022.1 6652 UTR 3 XP_005247079.1
XM_005247024.1 6652 UTR 3 XP_005247081.1
XM_005247027.1 6652 UTR 3 XP_005247084.1
XM_005247028.2 6652 UTR 3 XP_005247085.1
XM_006712605.1 6652 UTR 3 XP_006712668.1
XM_011511364.1 6652 UTR 3 XP_011509666.1
XM_011511365.1 6652 UTR 3 XP_011509667.1
XM_011511366.2 6652 UTR 3 XP_011509668.1
XM_011511367.1 6652 UTR 3 XP_011509669.1
XM_017004383.1 6652 UTR 3 XP_016859872.1
XM_017004384.1 6652 UTR 3 XP_016859873.1
XM_017004385.1 6652 UTR 3 XP_016859874.1
XM_017004386.1 6652 UTR 3 XP_016859875.1
XM_017004387.1 6652 UTR 3 XP_016859876.1
XM_017004388.1 6652 UTR 3 XP_016859877.1
XM_017004389.1 6652 UTR 3 XP_016859878.1
XM_017004390.1 6652 UTR 3 XP_016859879.1
XM_017004391.1 6652 UTR 3 XP_016859880.1

View Full Product Details