Product Details
- SNP ID
-
rs111782520
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240719045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGTCCTCACTGTACTCCACCTG[G/T]GCAGTGGCCAGGTTGAGCACGAACC
- Phenotype
-
MIM: 601255
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIF1A
PubMed Links
Gene Details
- Gene
- KIF1A
- Gene Name
- kinesin family member 1A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001244008.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1725A |
NP_001230937.1 |
NM_001320705.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1633A |
NP_001307634.1 |
XM_005247022.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1734A |
XP_005247079.1 |
XM_005247024.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1725A |
XP_005247081.1 |
XM_005247027.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1632A |
XP_005247084.1 |
XM_005247028.2 |
5033 |
Silent Mutation |
GCA,GCC |
A1624A |
XP_005247085.1 |
XM_006712605.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1716A |
XP_006712668.1 |
XM_011511364.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1734A |
XP_011509666.1 |
XM_011511365.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1642A |
XP_011509667.1 |
XM_011511366.2 |
5033 |
Silent Mutation |
GCA,GCC |
A1399A |
XP_011509668.1 |
XM_011511367.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1399A |
XP_011509669.1 |
XM_017004383.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1759A |
XP_016859872.1 |
XM_017004384.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1759A |
XP_016859873.1 |
XM_017004385.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1758A |
XP_016859874.1 |
XM_017004386.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1750A |
XP_016859875.1 |
XM_017004387.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1667A |
XP_016859876.1 |
XM_017004388.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1666A |
XP_016859877.1 |
XM_017004389.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1658A |
XP_016859878.1 |
XM_017004390.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1633A |
XP_016859879.1 |
XM_017004391.1 |
5033 |
Silent Mutation |
GCA,GCC |
A1733A |
XP_016859880.1 |
View Full Product Details