Product Details

SNP ID
rs111972561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:111979169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTGGCCTCAGCGTGCTGCTTGC[A/G]CGGATACCTCAGCGGGCGCCACGGT
Phenotype
MIM: 610298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD10 PubMed Links

Gene Details

Gene
ABHD10
Gene Name
abhydrolase domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272069.1 294 Silent Mutation GCA,GCG A36A NP_001258998.1
NM_018394.3 294 Silent Mutation GCA,GCG A36A NP_060864.1
XM_011512960.2 294 Silent Mutation GCA,GCG A36A XP_011511262.1
Gene
PHLDB2
Gene Name
pleckstrin homology like domain family B member 2
There are no transcripts associated with this gene.

View Full Product Details