Product Details

SNP ID
rs115637884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129096140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCTTTGCAGGCACAAACACTC[A/G]GGGTGTCTCTTTACCTAGTTCTCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISY1-RAB43 PubMed Links

Gene Details

Gene
ISY1-RAB43
Gene Name
ISY1-RAB43 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204890.1 Intron NP_001191819.1
Gene
RAB43
Gene Name
RAB43, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204883.1 Intron NP_001191812.1
NM_001204884.1 Intron NP_001191813.1
NM_001204885.1 Intron NP_001191814.1
NM_001204886.1 Intron NP_001191815.1
NM_001204887.1 Intron NP_001191816.1
NM_001204888.1 Intron NP_001191817.1
NM_198490.2 Intron NP_940892.1

View Full Product Details