Product Details

SNP ID
rs113807526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139358359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAAGAAAAGGATGATCAGAAT[G/T]AGGAAGTGGGAGATGATGTTTAAAA
Phenotype
MIM: 606990 MIM: 605810
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COPB2 PubMed Links

Gene Details

Gene
COPB2
Gene Name
coatomer protein complex subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004766.2 Intron NP_004757.1
XM_017007512.1 Intron XP_016863001.1
XM_017007513.1 Intron XP_016863002.1
Gene
MRPS22
Gene Name
mitochondrial ribosomal protein S22
There are no transcripts associated with this gene.

View Full Product Details