Product Details

SNP ID
rs115645244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142161582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTTTCAGGGAGCTTCCAGTCT[C/G]GTAGAGATTTACACAGAAGGCAGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GK5 PubMed Links

Gene Details

Gene
GK5
Gene Name
glycerol kinase 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039547.2 5782 UTR 3 NP_001034636.1
XM_011512624.2 5782 UTR 3 XP_011510926.1
XM_017006088.1 5782 Intron XP_016861577.1
XM_017006089.1 5782 UTR 3 XP_016861578.1
XM_017006090.1 5782 UTR 3 XP_016861579.1

View Full Product Details