Product Details

SNP ID
rs115476578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149319481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCCTAAGCATCTTTGTCTTA[C/G]CATCCCTCCAACCCAACATCTAGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TM4SF18 PubMed Links

Gene Details

Gene
TM4SF18
Gene Name
transmembrane 4 L six family member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184723.1 2693 UTR 3 NP_001171652.1
NM_138786.3 2693 UTR 3 NP_620141.1

View Full Product Details