Product Details

SNP ID
rs117453460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170092753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGACAAATATTCTCCAACTGTAA[G/T]AAAAGGTTCTCACCTTTTTCAGAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR160 PubMed Links

Gene Details

Gene
GPR160
Gene Name
G protein-coupled receptor 160
There are no transcripts associated with this gene.

Gene
PHC3
Gene Name
polyhomeotic homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308116.1 7319 UTR 3 NP_001295045.1
NM_024947.3 7319 UTR 3 NP_079223.3
XM_005247789.2 7319 UTR 3 XP_005247846.1
XM_005247791.3 7319 Intron XP_005247848.1
XM_005247793.2 7319 Intron XP_005247850.1
XM_006713753.1 7319 UTR 3 XP_006713816.1
XM_006713754.3 7319 UTR 3 XP_006713817.1
XM_006713756.2 7319 UTR 3 XP_006713819.1
XM_006713757.3 7319 Intron XP_006713820.1
XM_011513186.2 7319 UTR 3 XP_011511488.1
XM_011513187.2 7319 UTR 3 XP_011511489.1
XM_011513188.2 7319 UTR 3 XP_011511490.1
XM_011513189.2 7319 Intron XP_011511491.1
XM_017007235.1 7319 UTR 3 XP_016862724.1
XM_017007236.1 7319 UTR 3 XP_016862725.1
XM_017007237.1 7319 UTR 3 XP_016862726.1
XM_017007238.1 7319 UTR 3 XP_016862727.1
XM_017007239.1 7319 UTR 3 XP_016862728.1

View Full Product Details