Product Details

SNP ID
rs112061197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1692799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCAACAGCCCCAGTCAGTGAA[A/T]GAGGGTTCTTTCCTTTTTATTTTTG
Phenotype
MIM: 602422
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM53A PubMed Links

Gene Details

Gene
FAM53A
Gene Name
family with sequence similarity 53 member A
There are no transcripts associated with this gene.

Gene
SLBP
Gene Name
stem-loop binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306074.1 2164 UTR 3 NP_001293003.1
NM_001306075.1 2164 UTR 3 NP_001293004.1
NM_006527.3 2164 UTR 3 NP_006518.1

View Full Product Details