Product Details
- SNP ID
-
rs115233072
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1816898 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCACCAGTCTTTGAAAGTTCCTTC[C/T]TGATCTCCTGCAAGTCCTAATAAAA
- Phenotype
-
MIM: 134934
MIM: 604407
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FGFR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9328763] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FGFR3
- Gene Name
- fibroblast growth factor receptor 3
There are no transcripts associated with this gene.
- Gene
- LETM1
- Gene Name
- leucine zipper and EF-hand containing transmembrane protein 1
View Full Product Details