Product Details

SNP ID
rs114880853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1009993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGGCATCACTGGTCATAGCC[G/T]CAGTGCTCAATCAGCAAACATGGCC
Phenotype
MIM: 607852
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NKD2 PubMed Links

Gene Details

Gene
NKD2
Gene Name
naked cuticle homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271082.1 Intron NP_001258011.1
NM_033120.3 Intron NP_149111.1
XM_005248382.3 Intron XP_005248439.1
XM_005248383.3 Intron XP_005248440.1
XM_011514150.2 Intron XP_011512452.1
XM_017010000.1 Intron XP_016865489.1

View Full Product Details