Product Details

SNP ID
rs114930926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:7873421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTTGTTGTTGTGGTTTCTACC[A/G]CGGGCACCGGAGACCCACCCGACAC
Phenotype
MIM: 602568
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FASTKD3 PubMed Links

Gene Details

Gene
FASTKD3
Gene Name
FAST kinase domains 3
There are no transcripts associated with this gene.

Gene
MTRR
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002454.2 315 Missense Mutation ACG,GCG T60A NP_002445.2
NM_024010.2 315 Missense Mutation ACG,GCG T87A NP_076915.2

View Full Product Details