Product Details

SNP ID
rs115848010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78364341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTCATGCTTTATGCTTTTGCA[C/T]ATTAACATGATAATTGATTGAATAA
Phenotype
MIM: 606911
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCAMP1 PubMed Links

Gene Details

Gene
SCAMP1
Gene Name
secretory carrier membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290229.1 Intron NP_001277158.1
NM_004866.5 Intron NP_004857.4
XM_011543727.2 Intron XP_011542029.1
Gene
SCAMP1-AS1
Gene Name
SCAMP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details