Product Details
- SNP ID
-
rs115882637
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:80351764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACGTTCCAGCTCATTGAGGTCCC[A/G]GTTGACCGAATGTAAGTTGTCCTGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRSP8P
PubMed Links
Gene Details
- Gene
- CRSP8P
- Gene Name
- mediator complex subunit 27 pseudogene
There are no transcripts associated with this gene.
View Full Product Details