Product Details

SNP ID
rs115224400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:73517479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCAGAATTAGATGTCTTTCAC[C/T]TAAATGTCTCGGTGTTGACCAAAGG
Phenotype
MIM: 130590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1A1 PubMed Links

Gene Details

Gene
EEF1A1
Gene Name
eukaryotic translation elongation factor 1 alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001402.5 1783 UTR 3 NP_001393.1
XM_011535514.2 1783 Intron XP_011533816.1

View Full Product Details