Product Details

SNP ID
rs114803210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:88143122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCTATGAAAATGCAGGGCTAA[A/T]AAATTTTTAAGTATGAATGAATTCT
Phenotype
MIM: 114610
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CNR1 PubMed Links

Gene Details

Gene
CNR1
Gene Name
cannabinoid receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160226.1 2409 UTR 3 NP_001153698.1
NM_001160258.1 2409 UTR 3 NP_001153730.1
NM_001160259.1 2409 UTR 3 NP_001153731.1
NM_016083.4 2409 UTR 3 NP_057167.2
NM_033181.3 2409 UTR 3 NP_149421.2
XM_006715330.3 2409 UTR 3 XP_006715393.1
XM_011535424.1 2409 UTR 3 XP_011533726.1
XM_011535425.1 2409 UTR 3 XP_011533727.1
XM_011535426.1 2409 UTR 3 XP_011533728.1
XM_011535428.1 2409 UTR 3 XP_011533730.1
XM_017010238.1 2409 UTR 3 XP_016865727.1
XM_017010239.1 2409 UTR 3 XP_016865728.1
XM_017010240.1 2409 UTR 3 XP_016865729.1

View Full Product Details