Product Details

SNP ID
rs112217103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70861893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGTTATCTGTTTGAGAAAGTC[A/G]GATTCTTGCATCCCTGGCTGGGATC
Phenotype
MIM: 607497 MIM: 611372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GAT2 PubMed Links

Gene Details

Gene
B3GAT2
Gene Name
beta-1,3-glucuronyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080742.2 1478 Silent Mutation TCC,TCT S274S NP_542780.1
Gene
SMAP1
Gene Name
small ArfGAP 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044305.2 1478 UTR 3 NP_001037770.1
NM_001281439.1 1478 UTR 3 NP_001268368.1
NM_001281440.1 1478 UTR 3 NP_001268369.1
NM_021940.4 1478 UTR 3 NP_068759.2
XM_005248760.4 1478 Intron XP_005248817.1
XM_011536045.2 1478 Intron XP_011534347.1
XM_011536046.2 1478 Intron XP_011534348.1
XM_011536047.2 1478 Intron XP_011534349.1
XM_011536048.2 1478 UTR 3 XP_011534350.1
XM_017011170.1 1478 UTR 3 XP_016866659.1

View Full Product Details