Product Details

SNP ID
rs117923011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70863685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTCAATGTTTGATGAAAAATT[A/G]AGGTATGGAAAGACTCCACTGACTG
Phenotype
MIM: 607497 MIM: 611372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GAT2 PubMed Links

Gene Details

Gene
B3GAT2
Gene Name
beta-1,3-glucuronyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080742.2 Intron NP_542780.1
Gene
SMAP1
Gene Name
small ArfGAP 1
There are no transcripts associated with this gene.

View Full Product Details