Product Details

SNP ID
rs114480880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25966791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCCTCAAAGAGACGGATCAAG[A/G]AATGTCATGTGAGGAACACGGAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM38 PubMed Links

Gene Details

Gene
TRIM38
Gene Name
tripartite motif containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006355.4 858 Missense Mutation GAA,GGA E90G NP_006346.1
XM_005248799.4 858 Missense Mutation GAA,GGA E90G XP_005248856.1
XM_005248800.4 858 Missense Mutation GAA,GGA E90G XP_005248857.1
XM_017010175.1 858 Missense Mutation GAA,GGA E90G XP_016865664.1

View Full Product Details