Product Details

SNP ID
rs113012027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41737537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCACTAATTAATAATAATTGAT[A/C]ATAGATGACAATCTTCTATTTCATA
Phenotype
MIM: 169740 MIM: 600744
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PGC PubMed Links

Gene Details

Gene
PGC
Gene Name
progastricsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166424.1 Intron NP_001159896.1
NM_002630.3 Intron NP_002621.1
Gene
TFEB
Gene Name
transcription factor EB
There are no transcripts associated with this gene.

View Full Product Details