Product Details

SNP ID
rs116998188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42103505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCAGGTCCTTAATGGTTCCTT[C/G]TCTCTACCCTCCTTCCCCTCCCACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf132 PubMed Links

Gene Details

Gene
C6orf132
Gene Name
chromosome 6 open reading frame 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164446.2 4356 UTR 3 NP_001157918.1

View Full Product Details