Product Details

SNP ID
rs113210268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:485995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGCATGTGTAGTGACGCACGAC[A/G]AATTAAGAGTGAGTGAGAATGAAAG
Phenotype
MIM: 615329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC2 PubMed Links

Gene Details

Gene
EXOC2
Gene Name
exocyst complex component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018303.5 3685 UTR 3 NP_060773.3
XM_017011018.1 3685 UTR 3 XP_016866507.1
XM_017011019.1 3685 Intron XP_016866508.1
XM_017011020.1 3685 UTR 3 XP_016866509.1
XM_017011021.1 3685 UTR 3 XP_016866510.1
XM_017011022.1 3685 Intron XP_016866511.1
XM_017011023.1 3685 Intron XP_016866512.1
XM_017011024.1 3685 Intron XP_016866513.1
XM_017011025.1 3685 Intron XP_016866514.1
XM_017011026.1 3685 Intron XP_016866515.1

View Full Product Details