Product Details

SNP ID
rs116233809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:51617484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCTTGTGCCTCCAGAGTAAA[C/T]GTCTATAAACCAACTTCTTGCAAAT
Phenotype
MIM: 606702
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927082 PubMed Links

Gene Details

Gene
LOC101927082
Gene Name
uncharacterized LOC101927082
There are no transcripts associated with this gene.

Gene
PKHD1
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138694.3 13208 UTR 3 NP_619639.3
NM_170724.2 13208 Intron NP_733842.2
XM_011514680.2 13208 UTR 3 XP_011512982.1
XM_011514682.2 13208 UTR 3 XP_011512984.1
XM_011514683.2 13208 UTR 3 XP_011512985.1
XM_011514684.2 13208 UTR 3 XP_011512986.1
XM_011514685.1 13208 Intron XP_011512987.1
XM_011514686.2 13208 Intron XP_011512988.1
XM_011514687.1 13208 Intron XP_011512989.1
XM_011514688.2 13208 Intron XP_011512990.1
XM_011514690.2 13208 UTR 3 XP_011512992.1
XM_011514691.2 13208 UTR 3 XP_011512993.1
XM_017010944.1 13208 UTR 3 XP_016866433.1
XM_017010945.1 13208 UTR 3 XP_016866434.1
XM_017010946.1 13208 UTR 3 XP_016866435.1
XM_017010947.1 13208 UTR 3 XP_016866436.1
XM_017010948.1 13208 UTR 3 XP_016866437.1
XM_017010949.1 13208 UTR 3 XP_016866438.1
XM_017010950.1 13208 Intron XP_016866439.1
XM_017010951.1 13208 Intron XP_016866440.1
XM_017010952.1 13208 Intron XP_016866441.1

View Full Product Details