Product Details

SNP ID
rs113785065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57100788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGGAGAACCAGAAAAATGATCA[A/G]AATAATTCAGATACTAAGATCTCTG
Phenotype
MIM: 615708
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF451 PubMed Links

Gene Details

Gene
ZNF451
Gene Name
zinc finger protein 451
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031623.2 373 Intron NP_001026794.1
NM_001257273.1 373 Silent Mutation CAA,CAG Q124Q NP_001244202.1
NM_015555.2 373 Intron NP_056370.2
XM_005248994.1 373 Intron XP_005249051.1
XM_011514460.1 373 Intron XP_011512762.1
XM_011514461.1 373 Intron XP_011512763.1
XM_011514462.2 373 Intron XP_011512764.1
XM_011514463.2 373 Silent Mutation CAA,CAG Q96Q XP_011512765.1

View Full Product Details