Product Details

SNP ID
rs111379520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:41690335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGCAACAGTTCACTCCTCTCC[C/G]CCTTTAAGCCCACTTCCTCGGCCTC
Phenotype
MIM: 147290
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INHBA PubMed Links

Gene Details

Gene
INHBA
Gene Name
inhibin beta A subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012174.1 2603 Missense Mutation GCG,GGG A321G XP_016867663.1
XM_017012175.1 2603 Missense Mutation GCG,GGG A256G XP_016867664.1
XM_017012176.1 2603 Missense Mutation GCG,GGG A199G XP_016867665.1
Gene
INHBA-AS1
Gene Name
INHBA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details