Product Details

SNP ID
rs118005558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102456775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGGGACTGGACAGTGAGATAT[C/T]GGGAAACCCCATGAAAATAGATGCA
Phenotype
MIM: 613302 MIM: 615167 MIM: 610929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017621.3 10633 UTR 3 NP_060091.1
XM_005250464.2 10633 UTR 3 XP_005250521.1
XM_005250465.2 10633 UTR 3 XP_005250522.1
Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 10633 UTR 3 NP_001119812.1
NM_001271818.1 10633 UTR 3 NP_001258747.1
NM_001271819.1 10633 UTR 3 NP_001258748.1
NM_032831.3 10633 UTR 3 NP_116220.1

View Full Product Details