Product Details
- SNP ID
-
rs112473027
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:105884125 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTTTGTTTTCTGTGTCTTGCTT[A/G]AGAAGACCTTCACTTCCTCAAGGTC
- Phenotype
-
MIM: 615610
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN7L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150405061] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATXN7L1
- Gene Name
- ataxin 7 like 1
There are no transcripts associated with this gene.
- Gene
- CDHR3
- Gene Name
- cadherin related family member 3
View Full Product Details