Product Details

SNP ID
rs115553525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112483299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATTGACTTTACTAGAGTCCAGA[C/T]AGACATCACATGAGGATTTTTATCC
Phenotype
MIM: 603502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFRD1 PubMed Links

Gene Details

Gene
IFRD1
Gene Name
interferon related developmental regulator 1
There are no transcripts associated with this gene.

Gene
LSMEM1
Gene Name
leucine rich single-pass membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134468.1 532 Intron NP_001127940.1
NM_182597.2 532 Intron NP_872403.1
XM_011516074.1 532 UTR 5 XP_011514376.1
XM_011516075.2 532 Intron XP_011514377.1
XM_011516076.2 532 Intron XP_011514378.1
XM_017012028.1 532 UTR 5 XP_016867517.1

View Full Product Details