Product Details

SNP ID
rs116308113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112767153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAAATGGCAAGTTAGTGATAA[C/T]TGGTAGTATGAGTGCTTATTAATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM168 PubMed Links

Gene Details

Gene
TMEM168
Gene Name
transmembrane protein 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287497.1 2530 UTR 3 NP_001274426.1
NM_022484.5 2530 UTR 3 NP_071929.3
XM_017012523.1 2530 UTR 3 XP_016868012.1
XM_017012524.1 2530 Intron XP_016868013.1
XM_017012525.1 2530 UTR 3 XP_016868014.1

View Full Product Details