Product Details

SNP ID
rs115777530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128674723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGTCAGGCTGGTGGGAAATTA[C/T]ATTTCAGTCATCCCTGGGGCTTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM71F2 PubMed Links

Gene Details

Gene
FAM71F2
Gene Name
family with sequence similarity 71 member F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012454.5 Intron NP_001012457.3
NM_001128926.3 Intron NP_001122398.1
NM_001290254.1 Intron NP_001277183.1
NM_001290255.1 Intron NP_001277184.1
XM_011516128.2 Intron XP_011514430.1
XM_011516129.2 Intron XP_011514431.1
XM_011516130.2 Intron XP_011514432.1
XM_011516131.2 Intron XP_011514433.1
XM_011516132.2 Intron XP_011514434.1
XM_011516134.2 Intron XP_011514436.1
XM_011516135.2 Intron XP_011514437.1
XM_011516136.2 Intron XP_011514438.1
XM_017012147.1 Intron XP_016867636.1
XM_017012149.1 Intron XP_016867638.1

View Full Product Details