Product Details

SNP ID
rs114105891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139134760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTAGAGGAGCTACTTTCAAAA[A/G]GAGATTTCACTGGAGCTATTACCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC26 PubMed Links

Gene Details

Gene
TTC26
Gene Name
tetratricopeptide repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144920.2 223 Missense Mutation AGA,GGA R37G NP_001138392.1
NM_001144923.2 223 Missense Mutation AGA,GGA R37G NP_001138395.1
NM_001287512.1 223 Missense Mutation AGA,GGA R37G NP_001274441.1
NM_001287513.1 223 Intron NP_001274442.1
NM_001318333.1 223 UTR 5 NP_001305262.1
NM_001321740.1 223 Missense Mutation AGA,GGA R37G NP_001308669.1
NM_001321741.1 223 Missense Mutation AGA,GGA R37G NP_001308670.1
NM_001321742.1 223 Missense Mutation AGA,GGA R37G NP_001308671.1
NM_024926.3 223 Missense Mutation AGA,GGA R37G NP_079202.2

View Full Product Details