Product Details

SNP ID
rs115038293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34635019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAAGCATTCTTCAAATTACTCT[C/T]GTTCCCCAGCTGAGCTTTGTTGCTG
Phenotype
MIM: 601978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
There are no transcripts associated with this gene.

Gene
SIGMAR1
Gene Name
sigma non-opioid intracellular receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282205.1 1437 UTR 3 NP_001269134.1
NM_001282206.1 1437 UTR 3 NP_001269135.1
NM_001282207.1 1437 UTR 3 NP_001269136.1
NM_001282208.1 1437 UTR 3 NP_001269137.1
NM_001282209.1 1437 UTR 3 NP_001269138.1
NM_005866.3 1437 UTR 3 NP_005857.1
NM_147157.2 1437 UTR 3 NP_671513.1

View Full Product Details