Product Details

SNP ID
rs111705935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83622815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATGAGGATCCGGCCTGCCAGG[A/G]TGAGGATCCTCTGGAACCAAGAGGG
Phenotype
MIM: 611343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDNK PubMed Links

Gene Details

Gene
IDNK
Gene Name
IDNK, gluconokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001551.3 492 Intron NP_001001551.2
NM_001256915.1 492 Intron NP_001243844.1
XM_006717110.3 492 Intron XP_006717173.1
XM_006717111.3 492 Intron XP_006717174.1
XM_017014719.1 492 Intron XP_016870208.1
XM_017014720.1 492 Intron XP_016870209.1
XM_017014721.1 492 Missense Mutation GAT,GGT D16G XP_016870210.1
XM_017014722.1 492 Missense Mutation GAT,GGT D16G XP_016870211.1
XM_017014723.1 492 Missense Mutation GAT,GGT D16G XP_016870212.1
XM_017014724.1 492 Intron XP_016870213.1
XM_017014725.1 492 Intron XP_016870214.1

View Full Product Details