Product Details

SNP ID
rs111793097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96818178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATTCATTACATTCATAGGGCTT[A/C]TCCCCAGTGTGAGTTCTCTGATGCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF782 PubMed Links

Gene Details

Gene
ZNF782
Gene Name
zinc finger protein 782
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001662.1 2324 Missense Mutation GAG,GAT E615D NP_001001662.1
XM_005251742.4 2324 Missense Mutation GAG,GAT E615D XP_005251799.1
XM_005251744.3 2324 Missense Mutation GAG,GAT E591D XP_005251801.1
XM_011518315.2 2324 Missense Mutation GAG,GAT E615D XP_011516617.1
XM_011518318.2 2324 Missense Mutation GAG,GAT E615D XP_011516620.1
XM_011518320.2 2324 Missense Mutation GAG,GAT E609D XP_011516622.1
XM_017014343.1 2324 Missense Mutation GAG,GAT E609D XP_016869832.1
XM_017014344.1 2324 Missense Mutation GAG,GAT E483D XP_016869833.1

View Full Product Details