Product Details
- SNP ID
-
rs116352541
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:123381983 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAAGGGGTTGAGTGGTGGCTGTG[A/G]GAATGGGGTGGGTGTCCCTGCAGGG
- Phenotype
-
MIM: 609720
MIM: 613633
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRB2
PubMed Links
Gene Details
- Gene
- CRB2
- Gene Name
- crumbs 2, cell polarity complex component
There are no transcripts associated with this gene.
- Gene
- DENND1A
- Gene Name
- DENN domain containing 1A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020946.1 |
3589 |
Missense Mutation |
CCA,TCA |
P827S |
NP_065997.1 |
NM_024820.2 |
3589 |
Intron |
|
|
NP_079096.2 |
XM_005252109.2 |
3589 |
Missense Mutation |
CCA,TCA |
P888S |
XP_005252166.1 |
XM_005252111.4 |
3589 |
Missense Mutation |
CCA,TCA |
P842S |
XP_005252168.1 |
XM_005252113.4 |
3589 |
Intron |
|
|
XP_005252170.1 |
XM_006717195.3 |
3589 |
Missense Mutation |
CCA,TCA |
P885S |
XP_006717258.1 |
XM_011518882.2 |
3589 |
Missense Mutation |
CCA,TCA |
P903S |
XP_011517184.1 |
XM_011518883.2 |
3589 |
Missense Mutation |
CCA,TCA |
P884S |
XP_011517185.1 |
XM_011518884.2 |
3589 |
Missense Mutation |
CCA,TCA |
P874S |
XP_011517186.1 |
XM_011518885.2 |
3589 |
Missense Mutation |
CCA,TCA |
P867S |
XP_011517187.1 |
XM_011518886.2 |
3589 |
Missense Mutation |
CCA,TCA |
P860S |
XP_011517188.1 |
XM_011518887.2 |
3589 |
Intron |
|
|
XP_011517189.1 |
XM_017014947.1 |
3589 |
Missense Mutation |
CCA,TCA |
P876S |
XP_016870436.1 |
XM_017014948.1 |
3589 |
Missense Mutation |
CCA,TCA |
P870S |
XP_016870437.1 |
XM_017014949.1 |
3589 |
Missense Mutation |
CCA,TCA |
P861S |
XP_016870438.1 |
XM_017014950.1 |
3589 |
Missense Mutation |
CCA,TCA |
P843S |
XP_016870439.1 |
XM_017014951.1 |
3589 |
Missense Mutation |
CCA,TCA |
P838S |
XP_016870440.1 |
XM_017014952.1 |
3589 |
Missense Mutation |
CCA,TCA |
P629S |
XP_016870441.1 |
View Full Product Details