Product Details

SNP ID
rs117603246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126865600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTAGGTTCTCTGCAGAATGG[A/G]TCAGCAGTGTGAGAGGATGTCATTA
Phenotype
MIM: 611692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB34 PubMed Links

Gene Details

Gene
ZBTB34
Gene Name
zinc finger and BTB domain containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099270.1 Intron NP_001092740.1
XM_005251989.4 Intron XP_005252046.1
XM_011518699.2 Intron XP_011517001.1

View Full Product Details