Product Details

SNP ID
rs116070009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:290426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCCTGGAGGGCCCCCGCTTCC[A/G]GGCCTCCCAGTGCATCTATGCGCAT
Phenotype
MIM: 614757 MIM: 609650 MIM: 617032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFITM5 PubMed Links

Gene Details

Gene
IFITM5
Gene Name
interferon induced transmembrane protein 5
There are no transcripts associated with this gene.

Gene
NLRP6
Gene Name
NLR family pyrin domain containing 6
There are no transcripts associated with this gene.

Gene
PGGHG
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025092.4 1448 Missense Mutation CAG,CGG Q99R NP_079368.3
XM_011520383.2 1448 Missense Mutation CAG,CGG Q99R XP_011518685.1
XM_011520384.2 1448 Missense Mutation CAG,CGG Q99R XP_011518686.1
XM_011520385.2 1448 Missense Mutation CAG,CGG Q99R XP_011518687.1
XM_011520386.2 1448 Missense Mutation CAG,CGG Q99R XP_011518688.1
XM_011520387.2 1448 Missense Mutation CAG,CGG Q99R XP_011518689.1
XM_017018355.1 1448 Missense Mutation CAG,CGG Q99R XP_016873844.1

View Full Product Details