Product Details

SNP ID
rs114544023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5301019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGATTCTGAAATAAGGGCTCAAG[C/G]CCTACTCTGCCCAGAAAATAGGCGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B4 PubMed Links

Gene Details

Gene
OR51B4
Gene Name
olfactory receptor family 51 subfamily B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033179.2 928 Missense Mutation CCT,GCT P310A NP_149419.2

View Full Product Details