Product Details

SNP ID
rs116576673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21536562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATGGGTAAGATCAAGGTTTTC[A/G]GGGGGGAAATGGGAAATTTGTGTGG
Phenotype
MIM: 138571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GYS2 PubMed Links

Gene Details

Gene
GYS2
Gene Name
glycogen synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021957.3 2759 UTR 3 NP_068776.2
XM_006719063.3 2759 UTR 3 XP_006719126.1
XM_017019245.1 2759 Intron XP_016874734.1
Gene
SPX
Gene Name
spexin hormone
There are no transcripts associated with this gene.

View Full Product Details