Product Details
- SNP ID
-
rs117639846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:21537101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACTTCATCTTCCACATCACTGCT[C/G]TGAGGACTGGAGGCCTGAGACCCTG
- Phenotype
-
MIM: 138571
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GYS2
PubMed Links
Gene Details
- Gene
- GYS2
- Gene Name
- glycogen synthase 2
- Gene
- SPX
- Gene Name
- spexin hormone
There are no transcripts associated with this gene.
View Full Product Details