Product Details

SNP ID
rs112900131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11021817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAGTTTGCTCTGCTGTGTCCTA[A/G]GATTCCAAGTTGATGTGATTATACA
Phenotype
MIM: 168730 MIM: 613961 MIM: 612669
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 804 Intron NP_001278243.1
NM_001291315.1 804 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
PRH1-TAS2R14
Gene Name
PRH1-TAS2R14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316893.1 804 Intron NP_001303822.1
Gene
TAS2R19
Gene Name
taste 2 receptor member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176888.2 804 Missense Mutation NP_795369.1
Gene
TAS2R31
Gene Name
taste 2 receptor member 31
There are no transcripts associated with this gene.

View Full Product Details