Product Details

SNP ID
rs112265026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29797860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCACCAGAACGCAGTGGATAGG[C/T]GATTGGTATGTAGAGTCTAGTTCAG
Phenotype
MIM: 603213 MIM: 600999
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256269.1 Intron NP_001243198.1
NM_001256270.1 Intron NP_001243199.1
NM_007317.2 Intron NP_015556.1
Gene
MAZ
Gene Name
MYC associated zinc finger protein
There are no transcripts associated with this gene.

View Full Product Details