Product Details

SNP ID
rs114155397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3025788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCACCATGTGGGAAGTTTCTGGC[A/G]GCTGGCAACAATTACGGGCAGATTG
Phenotype
MIM: 617003 MIM: 615798 MIM: 615403 MIM: 605914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BICDL2 PubMed Links

Gene Details

Gene
BICDL2
Gene Name
BICD family like cargo adaptor 2
There are no transcripts associated with this gene.

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
There are no transcripts associated with this gene.

Gene
LOC100128770
Gene Name
uncharacterized LOC100128770
There are no transcripts associated with this gene.

Gene
THOC6
Gene Name
THO complex 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142350.1 416 Silent Mutation GCA,GCG A40A NP_001135822.1
NM_024339.3 416 Silent Mutation GCA,GCG A40A NP_077315.2
Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
There are no transcripts associated with this gene.

View Full Product Details