Product Details

SNP ID
rs116872004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30886776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATAAGTTAGACCCAACCAGAAG[G/T]GTGGCCAGGCATCCCAGGTGGAGGA
Phenotype
MIM: 605847 MIM: 600435
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL7C PubMed Links

Gene Details

Gene
BCL7C
Gene Name
BCL tumor suppressor 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286526.1 Intron NP_001273455.1
NM_004765.3 Intron NP_004756.2
XM_011545980.2 Intron XP_011544282.1
XM_017023885.1 Intron XP_016879374.1
XM_017023886.1 Intron XP_016879375.1
XM_017023887.1 Intron XP_016879376.1
XM_017023888.1 Intron XP_016879377.1
Gene
CTF1
Gene Name
cardiotrophin 1
There are no transcripts associated with this gene.

Gene
MIR762
Gene Name
microRNA 762
There are no transcripts associated with this gene.

Gene
MIR762HG
Gene Name
MIR762 host gene
There are no transcripts associated with this gene.

View Full Product Details