Product Details

SNP ID
rs112964318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3298985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCTAATGATTAGAACCAGAAG[C/T]TGAAGGCCCACTGAAGGAGTCCTTA
Phenotype
MIM: 612969 MIM: 604191 MIM: 601473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIGD7 PubMed Links

Gene Details

Gene
TIGD7
Gene Name
tigger transposable element derived 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033208.3 3253 Missense Mutation ACT,GCT T544A NP_149985.2
Gene
ZNF263
Gene Name
zinc finger protein 263
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005741.4 3253 Intron NP_005732.2
XM_006720831.1 3253 Intron XP_006720894.1
XM_006720832.1 3253 Intron XP_006720895.1
XM_006720833.1 3253 Intron XP_006720896.1
XM_006720834.3 3253 Intron XP_006720897.1
XM_011522344.1 3253 Intron XP_011520646.1
XM_017022850.1 3253 Intron XP_016878339.1
Gene
ZNF75A
Gene Name
zinc finger protein 75a
There are no transcripts associated with this gene.

View Full Product Details