Product Details
- SNP ID
-
rs111987725
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3658136 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCTGGCCCCCTGGCTGTCAGTGT[C/T]GCTCCAGGGCCTTGACAAGCAGCTC
- Phenotype
-
MIM: 125505
MIM: 606219
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNASE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2791] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNASE1
- Gene Name
- deoxyribonuclease I
- Gene
- TRAP1
- Gene Name
- TNF receptor associated protein 1
View Full Product Details