Product Details

SNP ID
rs117386203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:403665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCACTTTGCAAGTTTGTGTCAG[C/T]ACAAGTTCAGGACCAGCCTGAGCAA
Phenotype
MIM: 615839 MIM: 601818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DECR2 PubMed Links

Gene Details

Gene
DECR2
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020664.3 Intron NP_065715.1
Gene
NME4
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
There are no transcripts associated with this gene.

View Full Product Details