Product Details

SNP ID
rs114326454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67820853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTGCAGACTGGTCAGTTCTC[C/G]ATGGGTGGGCACCTGTCCCCATGGC
Phenotype
MIM: 611510 MIM: 607720
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPT PubMed Links

Gene Details

Gene
CENPT
Gene Name
centromere protein T
There are no transcripts associated with this gene.

Gene
TSNAXIP1
Gene Name
translin associated factor X interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288990.1 434 Silent Mutation TCC,TCG S54S NP_001275919.1
NM_001288991.1 434 Intron NP_001275920.1
NM_001288992.1 434 Intron NP_001275921.1
NM_001288993.1 434 Intron NP_001275922.1
NM_001288994.1 434 Intron NP_001275923.1
NM_018430.3 434 UTR 5 NP_060900.2
XM_005256049.4 434 Silent Mutation TCC,TCG S54S XP_005256106.1
XM_011523230.2 434 Silent Mutation TCC,TCG S54S XP_011521532.1
XM_011523231.2 434 Silent Mutation TCC,TCG S54S XP_011521533.1
XM_011523232.2 434 Silent Mutation TCC,TCG S54S XP_011521534.1
XM_011523233.2 434 Silent Mutation TCC,TCG S54S XP_011521535.1
XM_011523234.2 434 Silent Mutation TCC,TCG S54S XP_011521536.1
XM_011523235.1 434 UTR 5 XP_011521537.1
XM_011523236.2 434 Intron XP_011521538.1
XM_011523237.2 434 Intron XP_011521539.1
XM_011523238.1 434 Intron XP_011521540.1
XM_011523239.1 434 Silent Mutation TCC,TCG S54S XP_011521541.1
XM_011523240.1 434 Intron XP_011521542.1
XM_011523241.1 434 Intron XP_011521543.1
XM_011523242.1 434 Intron XP_011521544.1
XM_011523243.1 434 Intron XP_011521545.1
XM_017023445.1 434 Intron XP_016878934.1
XM_017023446.1 434 Intron XP_016878935.1
XM_017023447.1 434 Intron XP_016878936.1
XM_017023448.1 434 Intron XP_016878937.1
XM_017023449.1 434 Intron XP_016878938.1
XM_017023450.1 434 Intron XP_016878939.1
XM_017023451.1 434 Intron XP_016878940.1
XM_017023452.1 434 Intron XP_016878941.1
XM_017023453.1 434 Intron XP_016878942.1
XM_017023454.1 434 Intron XP_016878943.1
XM_017023455.1 434 Intron XP_016878944.1

View Full Product Details